This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05707510_g1
See other ETFA GE Assays ›
Gene Symbol:
Etfa
Gene Name:
electron transferring flavoprotein, alpha polypeptide
Gene Aliases:
2010200I21Rik, D9Ertd394e
Chromosome Location:
Chr.9: 55454436 - 55512243 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05707510_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Etfa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145615.4 NP_663590.3 2 - 3 362 84
GenBank mRNA AK028118.1 - 2 - 3 363 84
AK032830.1 - 2 - 3 315 84
AK133525.1 - 2 - 3 308 84
AK167572.1 - 2 - 3 335 84
AK168321.1 - 2 - 3 323 84
AK218235.1 - 1 - 2 53 84
BC003432.1 - 2 - 3 253 2646522 84
BC096645.1 - 2 - 3 221 1245213 84

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Etfa

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

transport
oxidation-reduction process
protein binding
electron carrier activity
oxidoreductase activity
flavin adenine dinucleotide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0