Assay Details
| Gene Symbol: Chfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289577.1 | NP_001276506.1 | 18 | 3069 | 104 | |
| NM_001289578.1 | NP_001276507.1 | 17 | 2853 | 104 | ||
| NM_001289579.1 | NP_001276508.1 | 19 | 3142 | 104 | ||
| NM_001289580.1 | NP_001276509.1 | 19 | 3236 | 104 | ||
| NM_172717.4 | NP_766305.2 | 18 | 3066 | 104 | ||
| GenBank mRNA | AK077629.1 | - | 18 | 3039 | 104 | |
| AK154038.1 | - | 18 | 3051 | 104 | ||
| AK155295.1 | - | 18 | 3040 | 104 | ||
| AK155525.1 | - | 18 | 3040 | 104 | ||
| AK169182.1 | - | 19 | 3093 | 104 | ||
| AK169193.1 | - | 18 | 3021 | 104 | ||
| BC049792.1 | - | 18 | 3023 | 6510480 | 104 | |
| CJ199205.1 | - | 1 | 277 | 104 | ||
More Information
Set Membership: |
|

Chfr
Genomic Map