This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05738792_s1
See other GDE1 GE Assays ›
Gene Symbol:
Gde1
Gene Name:
glycerophosphodiester phosphodiesterase 1
Gene Aliases:
1200003M13Rik, Mir16, RGS16
Chromosome Location:
Chr.7: 118688558 - 118705738 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05738792_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gde1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019580.4 NP_062526.1 6 1119 63
GenBank mRNA AK004583.1 - 6 1128 63
AK005361.1 - 6 1120 63
AK150807.1 - 6 1141 63
BC003902.1 - 6 1078 3601420 63

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gde1

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

glycerol metabolic process
lipid metabolic process
G-protein coupled receptor signaling pathway
phosphoric diester hydrolase activity
glycerophosphodiester phosphodiesterase activity
hydrolase activity
metal ion binding
glycerophosphoinositol glycerophosphodiesterase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline