This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05739120_s1
See other NUPR1 GE Assays ›
Gene Symbol:
Nupr1
Gene Name:
nuclear protein transcription regulator 1
Gene Aliases:
2310032H04Rik, Com1, p8
Chromosome Location:
Chr.7: 126623246 - 126625470 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05739120_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nupr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019738.1 NP_062712.1 3 405 78
GenBank mRNA AF131196.1 - 3 405 78
AK009572.1 - 3 425 78
BC002109.1 - 3 411 3487434 78

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nupr1

Gene Ontology Categories:

Function(s) Process(es)

acute inflammatory response
transcription, DNA-templated
regulation of transcription, DNA-templated
protein complex assembly
protein acetylation
male gonad development
response to toxic substance
cell growth
positive regulation of protein modification process
skeletal muscle cell differentiation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
positive regulation of apoptotic process
negative regulation of cell cycle
negative regulation of fibroblast proliferation
regulation of female gonad development
DNA binding
chromatin binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0