This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05763910_s1
See other COTL1 GE Assays ›
Gene Symbol:
Cotl1
Gene Name:
coactosin-like 1 (Dictyostelium)
Gene Aliases:
1810074P22Rik, 2010004C08Rik, Clp
Chromosome Location:
Chr.8: 119809214 - 119840579 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05763910_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cotl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028071.3 NP_082347.1 4 970 83
GenBank mRNA AK007994.1 - 4 932 83
AK008085.1 - 4 932 83
AK008597.1 - 1 397 83
AK010052.1 - 4 932 83
AK039644.1 - 4 933 83
AK148854.1 - 4 937 83
AK150888.1 - 4 932 83
AK151690.1 - 4 932 83
AK157694.1 - 4 933 83
AK172403.1 - 4 932 83
AK198264.1 - 1 177 83
BC010249.1 - 4 925 3155195 83
BC011068.1 - 5 873 3157387 83

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cotl1

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein cytoskeletal protein non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

defense response to fungus
actin binding
protein binding
enzyme binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0