This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05787098_g1
See other PPP4R2 GE Assays ›
Gene Symbol:
Ppp4r2
Gene Name:
protein phosphatase 4, regulatory subunit 2
Gene Aliases:
BE691708, C230060M08Rik
Chromosome Location:
Chr.6: 100833623 - 100869934 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05787098_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp4r2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_182939.4 NP_891984.1 2 - 3 371 107
GenBank mRNA AK082539.1 - 2 - 3 372 107
AK084169.1 - 2 - 3 332 107
BC056340.1 - 2 - 3 191 5703151 107
BC062117.1 - 1 - 2 45 107
BC098464.1 - 2 - 3 137 30093511 107
BC110429.1 - 2 - 3 376 5350332 107

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp4r2

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
mRNA processing
RNA splicing
regulation of double-strand break repair via homologous recombination
regulation of catalytic activity
protein binding
protein phosphatase type 4 regulator activity
protein binding, bridging

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0