This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05819436_s1
See other SRGAP2 GE Assays ›
Gene Symbol:
Srgap2
Gene Name:
SLIT-ROBO Rho GTPase activating protein 2
Gene Aliases:
9930124L22Rik, AI448945, FBP2, Fnbp2, srGAP3
Chromosome Location:
Chr.1: 131285251 - 131527369 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05819436_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Srgap2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081011.2 NP_001074480.2 23 6803 142
GenBank mRNA AK037136.1 - 1 142
BC151081.1 - 22 6421 9087994 142
BC151082.1 - 22 6422 9087995 142
BC158055.1 - 22 6419 9088163 142
BC172152.1 - 22 6419 9054822 142

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

lamellipodium assembly involved in ameboidal cell migration
signal transduction
nervous system development
extension of a leading process involved in cell motility in cerebral cortex radial glia guided migration
negative regulation of cell migration
substrate adhesion-dependent cell spreading
positive regulation of GTPase activity
filopodium assembly
neuron projection morphogenesis
actin filament severing
dendritic spine development
negative regulation of neuron migration
GTPase activator activity
protein binding
protein homodimerization activity
Rac GTPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0