This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05835189_s1
See other SSR1 GE Assays ›
Gene Symbol:
Ssr1
Gene Name:
signal sequence receptor, alpha
Gene Aliases:
2510001K09Rik, 6330400D04, AI159733, AI452176, SSR, TRAPA
Chromosome Location:
Chr.13: 37971401 - 37994203 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05835189_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ssr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025965.3 NP_080241.3 8 2402 104
GenBank mRNA AF326229.2 - 8 2378 104
AK010884.1 - 8 2349 104
AK013341.1 - 1 785 104
AK013910.1 - 1 1183 104
AK016687.1 - 1 513 104
AK031538.1 - 8 2383 104
AK166235.1 - 8 2372 104
BC011255.1 - 8 2333 4017600 104

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ssr1

Panther Classification:

Molecular Function -

membrane traffic protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f9vrx:80/100.66.79.34:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0