This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05878651_g1
See other EEF1A1 GE Assays ›
Gene Symbol:
Eef1a1
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 78478453 - 78481724 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05878651_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eef1a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010106.2 NP_034236.2 3 - 4 445 98
GenBank mRNA AK003188.1 - 3 - 4 394 98
AK011771.1 - 3 - 4 393 98
AK012341.1 - 3 - 4 394 98
AK032914.1 - 3 - 4 391 98
AK050635.1 - 3 - 4 392 98
AK076696.1 - 3 - 4 393 98
AK081725.1 - 3 - 4 393 98
AK083361.1 - 3 - 4 393 98
AK088041.1 - 3 - 4 394 98
AK135448.1 - 3 - 4 394 98
AK137223.1 - 3 - 4 393 98
AK141243.1 - 3 - 4 395 100
AK150798.1 - 3 - 4 393 98
AK151479.1 - 3 - 4 394 98
AK151773.1 - 3 - 4 393 98
AK152810.1 - 3 - 4 393 98
AK152993.1 - 3 - 4 393 98
AK153387.1 - 3 - 4 393 98
AK153402.1 - 3 - 4 393 98
AK153851.1 - 3 - 4 393 98
AK162049.1 - 3 - 4 393 98
AK162313.1 - 3 - 4 392 98
AK162440.1 - 3 - 4 397 98
AK163799.1 - 3 - 4 393 98
AK164113.1 - 3 - 4 395 98
AK164671.1 - 3 - 4 393 98
AK165938.1 - 3 - 4 393 98
AK166021.1 - 3 - 4 393 98
AK166033.1 - 3 - 4 393 98
AK166058.1 - 3 - 4 393 98
AK166089.1 - 3 - 4 393 98
AK166615.1 - 3 - 4 393 98
AK166807.1 - 3 - 4 393 98
AK167843.1 - 3 - 4 393 98
AK167873.1 - 3 - 4 392 98
AK168424.1 - 3 - 4 393 98
AK168467.1 - 3 - 4 399 98
AK200818.1 - 2 - 3 254 98
BC003969.1 - 2 - 3 363 98
BC004005.1 - 2 - 3 354 3492770 98
BC004067.1 - 2 - 3 382 3590965 98
BC005660.1 - 2 - 3 383 3588966 98
BC018223.1 - 3 - 4 381 4012653 98
BC018485.1 - 3 - 4 395 3491827 98
BC050124.1 - 1938 98
BC068258.1 - 2190 98
BC083069.1 - 3 - 4 381 6310448 98
BC092053.1 - 2 - 3 371 4008574 98
BC092276.1 - 2 - 3 376 3490007 98
BC092292.1 - 2 - 3 337 98
BC095965.1 - 2 - 3 371 6310404 98
BC108391.1 - 3 - 4 378 30242277 98
M22432.1 - 3 - 4 384 98

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eef1a1

Panther Classification:

Molecular Function -

translation factor

Gene Ontology Categories:

Function(s) Process(es)

translation
translational elongation
protein homooligomerization
cellular response to epidermal growth factor stimulus
tRNA binding
nucleotide binding
mRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0