This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05890123_g1
See other UBE2I GE Assays ›
Gene Symbol:
Ube2i
Gene Name:
ubiquitin-conjugating enzyme E2I
Gene Aliases:
5830467E05Rik, F830028O17Rik, UBC9, Ubce2i, Ubce9
Chromosome Location:
Chr.17: 25260510 - 25274311 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05890123_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube2i
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177609.1 NP_001171080.1 8 1485 99
NM_001177610.1 NP_001171081.1 7 774 99
NM_011665.4 NP_035795.1 7 812 99
GenBank mRNA AK003141.1 - 7 719 99
AK005058.1 - 7 794 99
AK011239.1 - 7 720 99
AK012282.1 - 7 738 99
AK088508.1 - 7 797 99
AK150575.1 - 7 755 99
AK160014.1 - 7 741 99
AK160988.1 - 7 738 99
AK165606.1 - 7 724 99
AK165615.1 - 7 742 99
AK166016.1 - 7 722 99
AK166657.1 - 8 1486 99
AK166930.1 - 7 718 99
AK167162.1 - 8 955 99
AK168212.1 - 7 741 99
AK168706.1 - 7 701 99
AK168766.1 - 7 735 99
U94402.1 - 7 808 99

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ube2i

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
cellular protein modification process
cell cycle
chromosome segregation
mitotic nuclear division
multicellular organism development
regulation of receptor activity
viral process
protein ubiquitination
protein sumoylation
positive regulation of intracellular steroid hormone receptor signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of transcription, DNA-templated
positive regulation of sequence-specific DNA binding transcription factor activity
cell division
positive regulation of SUMO transferase activity
nucleotide binding
ubiquitin-protein transferase activity
protein binding
ATP binding
protein C-terminus binding
transcription factor binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
SUMO transferase activity
enzyme binding
HLH domain binding
bHLH transcription factor binding
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
poly(A) RNA binding
SUMO conjugating enzyme activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity
RING-like zinc finger domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0