This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05899392_g1
See other HACD1 GE Assays ›
Gene Symbol:
Hacd1
Gene Name:
3-hydroxyacyl-CoA dehydratase 1
Gene Aliases:
Ptpla
Chromosome Location:
Chr.2: 14026366 - 14056035 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05899392_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hacd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012396.2 NP_001012396.2 2 383 86
NM_013935.3 NP_038963.3 2 383 86
GenBank mRNA AK004461.1 - 2 318 86
AK010119.1 - 2 228 86
AK076818.1 - 2 229 86
AK085898.1 - 384 86
BB662124.1 - 2 368 86
BC138116.1 - 3 371 8861135 86
BC141017.1 - 3 371 9055629 86

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Amplicon spans exons and probe does not span exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hacd1

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
fatty acid biosynthetic process
multicellular organism development
regulation of G2/M transition of mitotic cell cycle
positive regulation of cell-substrate adhesion
myotube differentiation
sphingolipid biosynthetic process
fatty acid elongation
cellular macromolecular complex assembly
very long-chain fatty acid biosynthetic process
cementum mineralization
regulation of G1/S transition of mitotic cell cycle
lyase activity
3-hydroxyacyl-CoA dehydratase activity
enzyme binding
hydroxyapatite binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0