This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05899710_g1
See other EEF1A1 GE Assays ›
Gene Symbol:
Eef1a1
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 78478453 - 78481724 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05899710_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eef1a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010106.2 NP_034236.2 4 476 122
GenBank mRNA AK003188.1 - 4 425 122
AK011771.1 - 4 424 122
AK012341.1 - 4 425 122
AK032914.1 - 4 422 122
AK050635.1 - 4 423 122
AK076696.1 - 4 424 122
AK081725.1 - 4 424 122
AK083361.1 - 4 424 122
AK088041.1 - 4 425 122
AK133725.1 - 4 122
AK135448.1 - 4 425 122
AK137223.1 - 4 424 122
AK150798.1 - 4 424 122
AK151479.1 - 4 425 122
AK151773.1 - 4 424 122
AK152810.1 - 4 424 122
AK152993.1 - 4 424 122
AK153387.1 - 4 424 122
AK153402.1 - 4 424 122
AK153851.1 - 4 424 122
AK162049.1 - 4 424 122
AK162313.1 - 4 423 122
AK162440.1 - 4 428 122
AK163799.1 - 4 424 122
AK164113.1 - 4 426 122
AK164671.1 - 4 424 122
AK165938.1 - 4 424 122
AK166021.1 - 4 424 122
AK166033.1 - 4 424 122
AK166058.1 - 4 424 122
AK166089.1 - 4 424 122
AK166615.1 - 4 424 122
AK166807.1 - 4 424 122
AK167843.1 - 4 424 122
AK167873.1 - 4 423 122
AK168424.1 - 4 424 122
AK168467.1 - 4 430 122
AK188863.1 - 2 122
AK200818.1 - 3 285 122
BC003969.1 - 3 394 122
BC004005.1 - 3 385 3492770 122
BC004067.1 - 3 413 3590965 122
BC005660.1 - 3 414 3588966 122
BC018223.1 - 4 412 4012653 122
BC018485.1 - 4 426 3491827 122
BC050124.1 - 1969 122
BC068258.1 - 2221 122
BC083069.1 - 4 412 6310448 122
BC092053.1 - 3 402 4008574 122
BC092276.1 - 3 407 3490007 122
BC092292.1 - 3 368 122
BC095965.1 - 3 402 6310404 122
BC108391.1 - 4 409 30242277 122
M22432.1 - 4 415 122
X13661.1 - 3 395 122

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Amplicon spans exons and probe does not span exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eef1a1

Panther Classification:

Molecular Function -

translation factor

Gene Ontology Categories:

Function(s) Process(es)

translation
translational elongation
protein homooligomerization
cellular response to epidermal growth factor stimulus
tRNA binding
nucleotide binding
mRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z47m5:80/100.66.77.18:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0