This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05901433_s1
See other ITM2C GE Assays ›
Gene Symbol:
Itm2c
Gene Name:
integral membrane protein 2C
Gene Aliases:
3110038L02Rik, BRI3, Bricd2c, E25, E25C, ITM3
Chromosome Location:
Chr.1: 85894510 - 85908698 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05901433_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Itm2c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022417.3 NP_071862.2 6 1049 99
GenBank mRNA AB030199.1 - 6 992 99
AF272044.1 - 6 956 99
AF282981.1 - 6 977 99
AK014144.1 - 2 225 99
AK036377.1 - 6 995 99
AK156930.1 - 6 952 99
AK197018.1 - 1 139 99
AK202940.1 - 1 139 99
BC012952.1 - 6 927 2648812 99

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Itm2c

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of neuron projection development
neuron differentiation
positive regulation of extrinsic apoptotic signaling pathway
beta-amyloid binding
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bjcbn:80/100.66.75.54:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0