This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05901966_s1
See other APBA1 GE Assays ›
Gene Symbol:
Apba1
Gene Name:
amyloid beta (A4) precursor protein binding, family A, member 1
Gene Aliases:
6430513E09Rik, Lin-10, Mint, Mint1, Urop11, X11, X11alpha
Chromosome Location:
Chr.19: 23758876 - 23949598 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05901966_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apba1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177034.3 NP_796008.2 13 6033 91
GenBank mRNA AK147583.1 - 13 6021 91
AK147623.1 - 13 5986 91

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Apba1

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
transport
intracellular protein transport
chemical synaptic transmission
locomotory behavior
regulation of gene expression
glutamate secretion
gamma-aminobutyric acid secretion
protein transport
multicellular organism growth
beta-amyloid binding
protein binding
phosphatidylinositol-4,5-bisphosphate binding
PDZ domain binding
protein complex binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0