This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05906008_s1
See other SLC7A14 GE Assays ›
Gene Symbol:
Slc7a14
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 14
Gene Aliases:
A930013N06, BC061928
Chromosome Location:
Chr.3: 31202854 - 31310574 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05906008_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172861.3 NP_766449.1 8 3135 78
GenBank mRNA AK173217.1 - 8 3111 78
BC061928.1 - 8 3111 5359765 78

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc7a14

Panther Classification:

Molecular Function -

amino acid transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
amino acid transport
negative regulation of phosphatase activity
L-alpha-amino acid transmembrane transport
molecular_function
amino acid transmembrane transporter activity
L-amino acid transmembrane transporter activity
antiporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0