This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05906047_s1
See other SLC7A11 GE Assays ›
Gene Symbol:
Slc7a11
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 11
Gene Aliases:
9930009M05Rik, AI451155, sut, xCT
Chromosome Location:
Chr.3: 50364936 - 50499087 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05906047_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011990.2 NP_036120.1 1 176 63
GenBank mRNA AB022345.1 - 1 143 63
AK037490.1 - 1 177 63
AK037742.1 - 1 151 63
AK076322.1 - 1 150 63
AK089202.1 - 1 149 63
AK089283.1 - 1 168 63
AK163168.1 - 1 150 63
AK165599.1 - 1 152 63
AK170568.1 - 1 45 63
AK171192.1 - 1 174 63
AK171252.1 - 1 150 63
AK171327.1 - 1 152 63
AY766236.1 - 1 143 63
AY766237.1 - 143 63
BB630675.1 - 1 177 63
BC141402.1 - 1 73 9056015 63
BC141408.1 - 1 73 9056021 63

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB022345.1, AK037742.1, AK089202.1, AK163168.1, AK165599.1, AK171192.1, AK171252.1, AY766236.1, AY766237.1, BC141402.1, BC141408.1, NM_011990.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc7a11

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
amino acid transport
response to toxic substance
neutral amino acid transport
platelet aggregation
modified amino acid transport
L-glutamate transmembrane transport
amino acid transmembrane transporter activity
cystine:glutamate antiporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0