This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05906456_g1
See other AIMP1 GE Assays ›
Gene Symbol:
Aimp1
Gene Name:
aminoacyl tRNA synthetase complex-interacting multifunctional protein 1
Gene Aliases:
9830137A06Rik, AIMP1/p43, EMAPII, Emap2, Scye1, p43
Chromosome Location:
Chr.3: 132660498 - 132683879 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05906456_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aimp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007926.2 NP_031952.2 7 996 100
GenBank mRNA AK087932.1 - 7 951 100
AK133859.1 - 7 949 100
BC002054.1 - 7 951 3496254 100
BY631162.1 - 2 283 100
U10118.1 - 7 947 100

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aimp1

Panther Classification:

Molecular Function -

translational protein

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
negative regulation of endothelial cell proliferation
carbohydrate metabolic process
glucose metabolic process
translation
apoptotic process
inflammatory response
cell adhesion
cell-cell signaling
leukocyte migration
tRNA binding
RNA binding
cytokine activity
protein homodimerization activity
GTPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0