This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05908260_s1
See other SYF2 GE Assays ›
Gene Symbol:
Syf2
Gene Name:
SYF2 homolog, RNA splicing factor (S. cerevisiae)
Gene Aliases:
1110018L13Rik, Cbpin, D4Bwg1551e, Gcipip, Ntc31, mp29, p29
Chromosome Location:
Chr.4: 134930980 - 134937537 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05908260_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Syf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026780.3 NP_081056.1 7 1088 84
GenBank mRNA AK003787.1 - 7 1090 84
AY033432.1 - 7 1089 84
BC051484.1 - 7 1082 1380946 84

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Syf2

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
mRNA processing
mitotic G2 DNA damage checkpoint
gastrulation
RNA splicing
embryonic organ development
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline