This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05910111_s1
See other NDNF GE Assays ›
Gene Symbol:
Ndnf
Gene Name:
neuron-derived neurotrophic factor
Gene Aliases:
A930038C07Rik, epidermacan
Chromosome Location:
Chr.6: 65671611 - 65712329 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05910111_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndnf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172399.3 NP_765987.2 4 1435 77
GenBank mRNA AK029210.1 - 4 1436 77
AK049802.1 - 4 1460 77
BC047154.1 - 4 1409 6489681 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndnf

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
nervous system development
positive regulation of cell-substrate adhesion
positive regulation of neuron projection development
cell growth
peptide cross-linking via chondroitin 4-sulfate glycosaminoglycan
extracellular matrix organization
negative regulation of neuron apoptotic process
glycosaminoglycan binding
heparin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6s775:80/100.66.79.102:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0