This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05911244_g1
See other RPS9 GE Assays ›
Gene Symbol:
Rps9
Gene Name:
ribosomal protein S9
Gene Aliases:
3010033P07Rik, AL022771, AL022885
Chromosome Location:
Chr.7: 3703953 - 3706899 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05911244_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rps9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029767.2 NP_084043.1 5 583 69
GenBank mRNA AK050591.1 - 5 586 69
AK081889.1 - 5 587 69
AK150800.1 - 5 581 69
AK150988.1 - 5 585 69
AK151029.1 - 5 581 69
AK151621.1 - 5 587 69
BC012491.1 - 4 1175 69
BC031746.1 - 4 573 3494314 69

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rps9

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

translation
regulation of translation
positive regulation of cell proliferation
positive regulation of translational fidelity
RNA binding
structural constituent of ribosome
rRNA binding
poly(A) RNA binding
translation regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline