This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05911812_s1
See other CCT2 GE Assays ›
Gene Symbol:
Cct2
Gene Name:
chaperonin containing Tcp1, subunit 2 (beta)
Gene Aliases:
Cctb
Chromosome Location:
Chr.10: 117050997 - 117063814 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05911812_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cct2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007636.2 NP_031662.2 16 1738 76
GenBank mRNA AA023539.1 - 1 70 76
AB041570.1 - 16 1569 76
AK012295.1 - 16 1698 76
AK075581.1 - 16 1711 76
AK088145.1 - 16 1700 76
AK145406.1 - 16 1739 76
AK166891.1 - 16 1711 76
BC007470.1 - 16 1702 3601569 76
BC026918.1 - 16 1704 4500896 76
Z31553.1 - 16 1689 76

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cct2

Panther Classification:

Molecular Function -

chaperonin

Gene Ontology Categories:

Function(s) Process(es)

protein folding
binding of sperm to zona pellucida
chaperone-mediated protein complex assembly
toxin transport
nucleotide binding
ATP binding
ubiquitin protein ligase binding
unfolded protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0