This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05912530_s1
See other UBE2L3 GE Assays ›
Gene Symbol:
Ube2l3
Gene Name:
ubiquitin-conjugating enzyme E2L 3
Gene Aliases:
C79827, UbcM4, Ubce7
Chromosome Location:
Chr.16: 17152015 - 17201498 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05912530_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube2l3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009456.2 NP_033482.1 4 1678 99
GenBank mRNA AJ130961.1 - 4 1667 99
AK165620.1 - 4 1660 99
AK167847.1 - 4 1667 99
BC049075.1 - 4 1665 99
X97042.1 - 4 1678 99

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ube2l3

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
regulation of transcription, DNA-templated
cell proliferation
protein ubiquitination
positive regulation of protein ubiquitination
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
cell cycle phase transition
positive regulation of ubiquitin-protein transferase activity
protein K11-linked ubiquitination
cellular response to steroid hormone stimulus
cellular response to glucocorticoid stimulus
positive regulation of protein targeting to mitochondrion
transcription coactivator activity
ubiquitin-protein transferase activity
ubiquitin protein ligase binding
poly(A) RNA binding
ubiquitin conjugating enzyme activity
ubiquitin-protein transferase activator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0