This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05912937_s1
See other GLCE GE Assays ›
Gene Symbol:
Glce
Gene Name:
glucuronyl C5-epimerase
Gene Aliases:
1110017N23Rik, AI747411, C130034A12Rik, Hsepi
Chromosome Location:
Chr.9: 62057248 - 62122720 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05912937_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glce
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_033320.4 NP_201577.3 2 712 96
GenBank mRNA AF325532.1 - 4 937 96
AF330049.1 - 2 946 96
BC094587.1 - 2 712 30618522 96

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Glce

Panther Classification:

Molecular Function -

isomerase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

glycosaminoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process
heparin biosynthetic process
isomerase activity
racemase and epimerase activity, acting on carbohydrates and derivatives
heparosan-N-sulfate-glucuronate 5-epimerase activity
UDP-glucuronate 5'-epimerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline