This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05914111_g1
See other PCNP GE Assays ›
Gene Symbol:
Pcnp
Gene Name:
PEST proteolytic signal containing nuclear protein
Gene Aliases:
1110018D06Rik, AI647035
Chromosome Location:
Chr.16: 56015506 - 56030177 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05914111_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pcnp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024622.2 NP_001019793.1 5 1908 97
GenBank mRNA AK150595.1 - 5 1869 97
AK152279.1 - 5 1872 97
AK159201.1 - 5 1879 97
AK168723.1 - 5 1834 97
AK172325.1 - 4 1912 97
BC023320.1 - 5 1876 97
BC106192.1 - 5 1817 97

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pcnp

Gene Ontology Categories:

Function(s)

cell cycle
protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline