This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05914670_s1
See other SEC22B GE Assays ›
Gene Symbol:
Sec22b
Gene Name:
SEC22 vesicle trafficking protein homolog B (S. cerevisiae)
Gene Aliases:
4930564D15Rik, AA517334, AI480645, C81333, ERS-24, Sec22l1
Chromosome Location:
Chr.3: 97901227 - 97922318 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05914670_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sec22b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011342.4 NP_035472.1 5 984 97
GenBank mRNA AF062527.1 - 1 62 97
AK017237.1 - 5 990 97
AK034973.1 - 5 996 97
AK037918.1 - 5 985 97
AK088514.1 - 5 984 97
AK089928.1 - 5 977 97
BC009024.1 - 5 946 3481731 97

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sec22b

Gene Ontology Categories:

Function(s) Process(es)

transport
ER to Golgi vesicle-mediated transport
protein transport
vesicle-mediated transport
regulation of organelle organization
positive regulation of protein catabolic process
vesicle fusion with Golgi apparatus
SNARE binding
SNAP receptor activity
syntaxin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0