This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05914858_s1
See other SLC16A9 GE Assays ›
Gene Symbol:
Slc16a9
Gene Name:
solute carrier family 16 (monocarboxylic acid transporters), member 9
Gene Aliases:
1200003C15Rik, 4930425B13Rik, AI552636
Chromosome Location:
Chr.10: 70245276 - 70285951 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05914858_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc16a9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025807.3 NP_080083.2 6 2017 95
GenBank mRNA AK004556.1 - 6 2018 95
AK004599.1 - 6 2196 95
AK004684.2 - 6 2075 95
BC034205.1 - 6 1973 4987055 95
BC055839.1 - 6 1975 4237811 95

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc16a9

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
monocarboxylic acid transport
ion transmembrane transport
urate metabolic process
transmembrane transport
anion transmembrane transport
organic acid transmembrane transport
molecular_function
monocarboxylic acid transmembrane transporter activity
symporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline