This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05914908_g1
See other EEF2 GE Assays ›
Gene Symbol:
Eef2
Gene Name:
eukaryotic translation elongation factor 2
Gene Aliases:
Ef-2
Chromosome Location:
Chr.10: 81176631 - 81182509 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05914908_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eef2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007907.2 NP_031933.1 14 2612 59
GenBank mRNA AK032997.1 - 14 2598 59
AK040474.1 - 14 2598 59
AK077866.1 - 14 2598 59
AK087985.1 - 14 2598 59
AK128976.1 - 14 2597 59
AK134195.1 - 14 2598 59
AK144874.1 - 14 2600 59
AK145509.1 - 14 2599 59
AK145545.1 - 14 2598 59
AK146508.1 - 14 2600 59
AK146947.1 - 14 2598 59
AK150136.1 - 14 2598 59
AK150902.1 - 14 2598 59
AK151487.1 - 14 2598 59
AK151812.1 - 14 2548 59
AK151945.1 - 14 2598 59
AK152146.1 - 14 2598 59
AK152447.1 - 14 2598 59
AK152587.1 - 14 2598 59
AK152826.1 - 14 2598 59
AK153275.1 - 14 2598 59
AK153936.1 - 14 2599 59
AK155325.1 - 14 2598 59
AK159155.1 - 14 2599 59
AK159421.1 - 14 2598 59
AK159514.1 - 14 2598 59
AK159806.1 - 14 2598 59
AK159829.1 - 14 2598 59
AK161993.1 - 7 1198 59
AK163063.1 - 14 2598 59
AK164406.1 - 6 1044 59
AK166212.1 - 14 2598 59
AK166595.1 - 14 59
AK166596.1 - 14 2598 59
AK166719.1 - 14 2600 59
AK166747.1 - 14 2598 59
AK166751.1 - 14 2596 59
AK166758.1 - 14 2599 59
AK166788.1 - 14 2600 59
AK166815.1 - 14 2599 59
AK166829.1 - 14 2599 59
AK166851.1 - 14 2596 59
AK166968.1 - 14 2598 59
AK167081.1 - 14 2599 59
AK167093.1 - 14 2598 59
AK167109.1 - 14 2598 59
AK167115.1 - 14 2598 59
AK167199.1 - 14 2598 59
AK167208.1 - 14 2596 59
AK167221.1 - 14 2598 59
AK167230.1 - 14 2598 59
AK167281.1 - 14 2598 59
AK168527.1 - 14 2599 59
AK168615.1 - 14 2599 59
AK168638.1 - 14 2599 59
AK168659.1 - 14 2599 59
AK168794.1 - 14 2598 59
AK168827.1 - 14 2598 59
AK168874.1 - 14 2591 59
AK169013.1 - 14 2598 59
AK189877.1 - 1 29 59
AK189936.1 - 1 29 59
AK190628.1 - 1 29 59
AK190763.1 - 1 29 59
AK190861.1 - 1 29 59
AK190927.1 - 1 29 59
AK191362.1 - 1 29 59
AK191366.1 - 1 29 59
AK191589.1 - 160 59
AK191622.1 - 1 29 59
AK191649.1 - 1 29 59
AK191810.1 - 1 29 59
AK191929.1 - 1 29 59
AK192150.1 - 1 29 59
AK192424.1 - 1 29 59
AK192757.1 - 1 29 59
AK192765.1 - 1 29 59
AK193267.1 - 1 29 59
AK193334.1 - 1 59
AK193414.1 - 1 29 59
AK193679.1 - 1 29 59
AK193930.1 - 1 29 59
AK193934.1 - 1 60
AK193935.1 - 1 29 60
AK193939.1 - 1 29 59
AK195004.1 - 1 29 59
AK195059.1 - 1 29 59
AK195098.1 - 1 29 59
AK195214.1 - 1 29 59
AK195454.1 - 1 29 59
AK195506.1 - 1 29 59
AK196079.1 - 1 29 59
AK196233.1 - 1 29 59
AK196287.1 - 1 29 59
AK196489.1 - 1 29 59
AK196556.1 - 1 29 59
AK196849.1 - 1 29 59
AK196962.1 - 1 29 59
AK197206.1 - 167 59
AK197222.1 - 1 29 59
AK197244.1 - 1 29 59
AK197312.1 - 1 59
AK197355.1 - 1 29 59
AK197363.1 - 1 29 59
AK197618.1 - 1 29 59
AK197848.1 - 1 59
AK198166.1 - 1 29 59
AK198483.1 - 1 29 59
AK198569.1 - 1 29 59
AK198683.1 - 1 29 59
AK199179.1 - 1 29 59
AK199520.1 - 1 29 59
AK199588.1 - 1 29 59
AK199703.1 - 1 29 59
AK199709.1 - 1 29 59
AK199751.1 - 1 29 59
AK200092.1 - 1 29 59
AK200307.1 - 1 29 59
AK200465.1 - 1 29 59
AK200510.1 - 1 29 59
AK200525.1 - 1 29 59
AK201052.1 - 1 29 59
AK201117.1 - 1 29 59
AK201643.1 - 1 29 59
AK202077.1 - 1 29 59
AK202127.1 - 1 29 59
AK202326.1 - 1 29 59
AK202505.1 - 1 29 59
AK202866.1 - 1 59
AK203693.1 - 1 29 59
AK204680.1 - 1 29 59
AK204768.1 - 1 29 59
AK207046.1 - 1 29 59
AK211931.1 - 1 29 59
BC002233.1 - 4 804 59
BC007152.1 - 14 2596 3600352 59
BC013263.1 - 14 2584 59
BC060707.1 - 13 2473 59

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eef2

Panther Classification:

Molecular Function -

translation elongation factor

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
translation
translational elongation
positive regulation of translation
nucleotide binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0