This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05917903_s1
See other NENF GE Assays ›
Gene Symbol:
Nenf
Gene Name:
neuron derived neurotrophic factor
Gene Aliases:
1110060M21Rik, SCIRP10, Spuf
Chromosome Location:
Chr.1: 191306797 - 191318118 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05917903_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nenf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025424.2 NP_079700.1 4 543 64
GenBank mRNA AK002413.1 - 4 544 64
AK004329.1 - 4 532 64
AK009304.1 - 4 534 64
AK020403.1 - 4 525 64
AK155319.1 - 4 542 64
BC048464.1 - 4 531 6596336 64

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nenf

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of appetite
positive regulation of MAPK cascade
growth factor activity
heme binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0