This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm06653751_g1
See other EEF1A1 GE Assays ›
Gene Symbol:
Eef1a1
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 78478453 - 78481724 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm06653751_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eef1a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010106.2 NP_034236.2 8 1740 103
GenBank mRNA AK003188.1 - 8 1676 103
AK012341.1 - 8 1689 103
AK032914.1 - 8 1686 103
AK050635.1 - 8 1689 103
AK076696.1 - 8 1689 103
AK081725.1 - 8 1688 103
AK083361.1 - 8 1688 103
AK088041.1 - 8 1687 103
AK135448.1 - 8 1689 103
AK137223.1 - 8 1688 103
AK141243.1 - 8 1690 103
AK150798.1 - 8 1688 103
AK151479.1 - 8 1689 103
AK151773.1 - 8 1688 103
AK152810.1 - 8 1688 103
AK153851.1 - 8 1690 103
AK162049.1 - 8 1688 103
AK162313.1 - 8 1687 103
AK162440.1 - 8 1692 103
AK163799.1 - 8 1689 103
AK164113.1 - 8 1690 103
AK165938.1 - 8 1688 103
AK166021.1 - 8 1688 103
AK166033.1 - 8 1688 103
AK166058.1 - 8 1688 103
AK166089.1 - 8 1688 103
AK166807.1 - 8 1688 103
AK167843.1 - 8 1688 103
AK167873.1 - 8 1686 103
AK168424.1 - 8 1690 103
AK168467.1 - 8 1696 103
AK199391.1 - 1 320 103
AK204572.1 - 1 318 103
AK206186.1 - 1 281 103
AK210662.1 - 1 327 104
BC003969.1 - 7 1658 103
BC004005.1 - 7 1648 3492770 103
BC005660.1 - 7 1677 3588966 103
BC018223.1 - 8 1675 4012653 103
BC018485.1 - 8 1689 3491827 103
BC040689.1 - 5 1180 103
BC050124.1 - 3233 103
BC068258.1 - 3485 103
BC083069.1 - 8 1676 6310448 103
BC092053.1 - 7 1665 4008574 103
BC092276.1 - 7 1671 3490007 103
BC092292.1 - 7 1630 103
BC095965.1 - 7 1666 6310404 103
BC099609.1 - 2 558 103
BC108391.1 - 8 1673 30242277 103
X13661.1 - 7 1657 103

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eef1a1

Panther Classification:

Molecular Function -

translation factor

Gene Ontology Categories:

Function(s) Process(es)

translation
translational elongation
protein homooligomerization
cellular response to epidermal growth factor stimulus
tRNA binding
nucleotide binding
mRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0