This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07293101_gH
See other ALDOA GE Assays ›
Gene Symbol:
Aldoa
Gene Name:
aldolase A, fructose-bisphosphate
Gene Aliases:
Aldo-1, Aldo1
Chromosome Location:
Chr.7: 126795234 - 126809313 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07293101_gH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aldoa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177307.1 NP_001170778.1 2 - 3 284 109
GenBank mRNA CF617557.1 - 1 - 2 43 109
EF662059.1 - 1 - 2 138 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

aldolase

Gene Ontology Categories:

Function(s) Process(es)

fructose metabolic process
glycolytic process
ATP biosynthetic process
striated muscle contraction
regulation of cell shape
fructose 1,6-bisphosphate metabolic process
muscle cell cellular homeostasis
fructose-bisphosphate aldolase activity
cytoskeletal protein binding
poly(A) RNA binding
fructose binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0