This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07293759_m1
See other ECHDC2 GE Assays ›
Gene Symbol:
Echdc2
Gene Name:
enoyl Coenzyme A hydratase domain containing 2
Gene Aliases:
1300017C12Rik, 2610009M20Rik, D4Ertd765e
Chromosome Location:
Chr.4: 108165436 - 108179308 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07293759_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Echdc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001254754.1 NP_001241683.1 6 - 7 739 95
NM_026728.4 NP_081004.2 7 - 8 807 95
GenBank mRNA AK005030.1 - 7 - 8 748 95
AK011365.1 - 6 - 7 685 95
AK153878.1 - 7 - 8 782 95
AK160482.1 - 5 - 6 510 95
AK166388.1 - 7 - 8 779 95
AK166965.1 - 7 - 8 749 95
BC025104.1 - 7 - 8 753 3591648 95
BC025454.1 - 3 - 4 291 5352625 95

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Echdc2

Panther Classification:

Molecular Function -

hydratase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
metabolic process
catalytic activity
lyase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0