This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07294003_m1
See other GJA5 GE Assays ›
Gene Symbol:
Gja5
Gene Name:
gap junction protein, alpha 5
Gene Aliases:
5730555N10Rik, Cnx40, Cx40, Gja-5
Chromosome Location:
Chr.3: 97032402 - 97053634 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07294003_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gja5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271628.1 NP_001258557.1 2 - 3 145 87
GenBank mRNA AY390401.1 - 2 - 3 145 87

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AY390401.1, NM_001271628.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
blood vessel development
renal system process involved in regulation of systemic arterial blood pressure
regulation of systemic arterial blood pressure
negative regulation of glomerular filtration
outflow tract morphogenesis
cardiac conduction system development
mitral valve development
pulmonary valve formation
ventricular septum development
atrial septum development
septum primum development
atrial ventricular junction remodeling
potassium ion transport
cell communication
cell communication by chemical coupling
cell communication by electrical coupling
positive regulation of cell communication by chemical coupling
gap junction assembly
embryonic limb morphogenesis
embryonic heart tube development
foramen ovale closure
regulation of vasodilation
negative regulation of blood pressure
positive regulation of vasoconstriction
positive regulation of vasodilation
artery morphogenesis
protein oligomerization
transmembrane transport
regulation of cardiac muscle contraction
regulation of ventricular cardiac muscle cell membrane repolarization
regulation of atrial cardiac muscle cell membrane depolarization
regulation of ventricular cardiac muscle cell membrane depolarization
ventricular septum morphogenesis
atrial septum morphogenesis
cardiac conduction
ventricular cardiac muscle cell action potential
SA node cell action potential
AV node cell to bundle of His cell communication by electrical coupling
bundle of His cell to Purkinje myocyte communication by electrical coupling
cell communication by electrical coupling involved in cardiac conduction
cell communication involved in cardiac conduction
AV node cell to bundle of His cell communication
regulation of heart rate by cardiac conduction
regulation of AV node cell action potential
regulation of bundle of His cell action potential
regulation of Purkinje myocyte action potential
regulation of atrial cardiac muscle cell action potential
regulation of renin secretion into blood stream
regulation of membrane depolarization during cardiac muscle cell action potential
vasomotion
gap junction channel activity
gap junction hemi-channel activity
connexin binding
gap junction channel activity involved in cardiac conduction electrical coupling
gap junction channel activity involved in AV node cell-bundle of His cell electrical coupling

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline