This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07294055_g1
See other ARC GE Assays ›
Gene Symbol:
Arc
Gene Name:
activity regulated cytoskeletal-associated protein
Gene Aliases:
Arc3.1, C86064, arg3.1, mArc
Chromosome Location:
Chr.15: 74669081 - 74672570 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07294055_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001276684.1 NP_001263613.1 2 - 3 2132 70
NM_018790.3 NP_061260.1 2 - 3 2135 70
GenBank mRNA AF162777.1 - 2 - 3 2126 70
BC023127.1 - 2 - 3 2121 3498057 70

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arc

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
cytoskeleton organization
multicellular organism development
endoderm development
anterior/posterior pattern specification
cell migration
regulation of cell morphogenesis
positive regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
actin binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0