Assay Details
| Gene Symbol: Ctla4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281976.1 | NP_001268905.1 | 2 - 3 | 602 | 100 | |
| GenBank mRNA | U90270.1 | - | 1 - 2 | 66 | 100 | |
More Information
Set Membership: |
|

Ctla4
Genomic Map