This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07295136_m1
See other IL7 GE Assays ›
Gene Symbol:
Il7
Gene Name:
interleukin 7
Gene Aliases:
A630026I06Rik, Il-7, hlb368
Chromosome Location:
Chr.3: 7572028 - 7613427 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07295136_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008371.4 NP_032397.1 4 - 5 597 72
GenBank mRNA AK040399.1 - 4 - 5 381 72
AK041307.1 - 4 - 5 598 72
AK041403.1 - 299 72
AK052452.1 - 5 - 6 1285 72
AK139858.1 - 5 - 6 958 72
AK145184.1 - 4 - 5 888 72
BC110553.2 - 4 - 5 506 40047584 72
X07962.1 - 4 - 5 897 72

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Il7

Gene Ontology Categories:

Function(s) Process(es)

T cell lineage commitment
immune response
humoral immune response
cell-cell signaling
organ morphogenesis
regulation of gene expression
positive regulation of B cell proliferation
negative regulation of catalytic activity
bone resorption
positive regulation of T cell differentiation
positive regulation of organ growth
homeostasis of number of cells within a tissue
negative regulation of extrinsic apoptotic signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
cytokine activity
cytokine receptor binding
interleukin-7 receptor binding
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0