Assay Details
| Gene Symbol: 2810408A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027419.3 | NP_081695.2 | 1 - 2 | 681 | 88 | |
| GenBank mRNA | AK013042.1 | - | 1 - 2 | 682 | 88 | |
| AK160144.1 | - | 1 - 2 | 591 | 89 | ||
| BC051546.1 | - | 1 - 2 | 642 | 5255109 | 88 | |
| BC069874.1 | - | 1 - 2 | 681 | 4216824 | 88 | |
More Information
Set Membership: |
|

2810408A11Rik
Genomic Map