Assay Details
| Gene Symbol: 2810408A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK160144.1 | - | 7 - 8 | 1294 | 106 | |
| AK171674.1 | - | 7 - 8 | 1446 | 106 | ||
| BC051546.1 | - | 7 - 8 | 1361 | 5255109 | 106 | |
More Information
Set Membership: |
|

2810408A11Rik
Genomic Map