This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07295489_g1
See other ALDOA GE Assays ›
Gene Symbol:
Aldoa
Gene Name:
aldolase A, fructose-bisphosphate
Gene Aliases:
Aldo-1, Aldo1
Chromosome Location:
Chr.7: 126795234 - 126809313 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07295489_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aldoa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007438.4 NP_031464.1 1 - 2 209 116
GenBank mRNA AK002561.1 - 1 - 2 98 116
AK145639.1 - 1 - 2 96 116
AK162290.1 - 1 - 2 97 116
BC043026.1 - 1 - 2 85 5691975 116
BC050896.1 - 1 - 2 127 5715436 116
BC066218.1 - 1 - 2 105 30054842 116
BC066801.1 - 1 - 2 105 30072521 116
BC089495.1 - 1 - 2 138 30312668 116
CN694938.1 - 1 - 2 210 116
EF662060.1 - 1 - 2 90 116

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK145639.1, BC043026.1, BC050896.1, BC066218.1, BC066801.1, BC089495.1, EF662060.1, NM_007438.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aldoa

Panther Classification:

Molecular Function -

aldolase

Gene Ontology Categories:

Function(s) Process(es)

fructose metabolic process
glycolytic process
ATP biosynthetic process
striated muscle contraction
regulation of cell shape
fructose 1,6-bisphosphate metabolic process
muscle cell cellular homeostasis
fructose-bisphosphate aldolase activity
cytoskeletal protein binding
poly(A) RNA binding
fructose binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0