This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07295722_m1
See other GPX4 GE Assays ›
Gene Symbol:
Gpx4
Gene Name:
glutathione peroxidase 4
Gene Aliases:
GPx-4, GSHPx-4, PHGPx, mtPHGPx, snGPx
Chromosome Location:
Chr.10: 80047166 - 80056439 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07295722_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpx4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037741.3 NP_001032830.2 2 - 3 373 92
NM_008162.3 NP_032188.3 2 - 3 319 92
GenBank mRNA AB072797.1 - 2 - 3 355 92
AF045768.1 - 2 - 3 237 92
AF045769.1 - 2 - 3 128 92
AF274027.1 - 2 - 3 349 92
AK006441.1 - 2 - 3 214 92
AK160389.1 - 2 - 3 146 92
AK160509.1 - 2 - 3 122 92
AK167881.1 - 2 - 3 223 92
AK168027.1 - 2 - 3 146 92
BC083137.1 - 1 - 2 106 92
BC106147.1 - 2 - 3 166 4936866 92
BU961981.1 - 376 92
BY238083.1 - 2 - 3 243 92
CA466677.1 - 2 - 3 300 92
D87896.1 - 2 - 3 319 92

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpx4

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

chromatin organization
glutathione metabolic process
response to oxidative stress
multicellular organism development
spermatogenesis
hydrogen peroxide catabolic process
regulation of inflammatory response
oxidation-reduction process
peroxidase activity
glutathione peroxidase activity
selenium binding
oxidoreductase activity
glutathione binding
phospholipid-hydroperoxide glutathione peroxidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0