This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07295723_m1
See other GPX4 GE Assays ›
Gene Symbol:
Gpx4
Gene Name:
glutathione peroxidase 4
Gene Aliases:
GPx-4, GSHPx-4, PHGPx, mtPHGPx, snGPx
Chromosome Location:
Chr.10: 80047166 - 80056439 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07295723_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpx4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037741.3 NP_001032830.2 3 - 4 522 67
NM_008162.3 NP_032188.3 3 - 4 468 67
GenBank mRNA AB072797.1 - 3 - 4 504 67
AF045768.1 - 3 - 4 386 67
AF045769.1 - 3 - 4 277 67
AF274027.1 - 3 - 4 498 67
AK006441.1 - 3 - 4 363 67
AK160389.1 - 3 - 4 295 67
AK160509.1 - 3 - 4 271 67
AK168027.1 - 3 - 4 295 67
BC083137.1 - 2 - 3 255 67
BC106147.1 - 3 - 4 315 4936866 67
CA466677.1 - 3 - 4 449 67
D87896.1 - 3 - 4 468 67

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpx4

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

chromatin organization
glutathione metabolic process
response to oxidative stress
multicellular organism development
spermatogenesis
hydrogen peroxide catabolic process
regulation of inflammatory response
oxidation-reduction process
peroxidase activity
glutathione peroxidase activity
selenium binding
oxidoreductase activity
glutathione binding
phospholipid-hydroperoxide glutathione peroxidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0