This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07297014_m1
See other SLC25A17 GE Assays ›
Gene Symbol:
Slc25a17
Gene Name:
solute carrier family 25 (mitochondrial carrier, peroxisomal membrane protein), member 17
Gene Aliases:
34kDa, 47kDa, PMP34, Pmp47
Chromosome Location:
Chr.15: 81318921 - 81360765 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07297014_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc25a17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011399.3 NP_035529.1 3 - 4 222 102
GenBank mRNA AJ006341.1 - 3 - 4 234 102
AK002388.1 - 3 - 4 223 102
AK003672.1 - 3 - 4 209 102
BC008571.1 - 3 - 4 224 3587806 102
BC011292.1 - 3 - 4 204 3962111 102

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc25a17

Gene Ontology Categories:

Function(s) Process(es)

fatty acid beta-oxidation
transport
ADP transport
ATP transport
fatty acid transport
coenzyme A transmembrane transport
FAD transmembrane transport
NAD transport
transmembrane transport
AMP transport
cation transmembrane transport
nucleotide transmembrane transport
ATP transmembrane transporter activity
ADP transmembrane transporter activity
coenzyme A transmembrane transporter activity
FAD transmembrane transporter activity
FMN transmembrane transporter activity
chaperone binding
NAD transporter activity
AMP transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0