This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07297311_g1
See other ATP5L GE Assays ›
Gene Symbol:
Atp5l
Gene Name:
ATP synthase, H+ transporting, mitochondrial F0 complex, subunit G
Gene Aliases:
4933437C06Rik
Chromosome Location:
Chr.9: 44913248 - 44920742 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07297311_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013795.5 NP_038823.2 2 - 3 295 110
GenBank mRNA AK005175.1 - 2 - 3 242 110
AK005321.1 - 2 - 3 272 110
AK005380.1 - 2 - 3 273 110
AK008002.1 - 2 - 3 232 110
AK008012.1 - 2 - 3 269 110
AK008030.1 - 2 - 3 258 110
AK008036.1 - 2 - 3 272 110
AK008087.1 - 2 - 3 246 110
AK008169.1 - 2 - 3 272 110
AK008183.1 - 2 - 3 260 110
AK008251.1 - 2 - 3 260 110
AK008321.1 - 2 - 3 268 110
AK009465.1 - 2 - 3 261 110
AK009468.1 - 2 - 3 261 110
AK009471.1 - 2 - 3 261 110
AK009612.1 - 2 - 3 254 110
AK009806.1 - 2 - 3 260 110
AK011038.1 - 2 - 3 239 110
AK011046.1 - 2 - 3 239 110
AK011842.1 - 2 - 3 232 110
AK019252.1 - 2 - 3 261 110
AK019274.1 - 2 - 3 261 110
AK019320.1 - 2 - 3 268 110
AK019343.1 - 2 - 3 232 110
AK019350.1 - 2 - 3 260 110
AK019355.1 - 2 - 3 260 110
BC031384.1 - 2 - 3 268 5356491 110
BC086880.1 - 2 - 3 231 6762989 110
BC089555.1 - 2 - 3 271 30312783 110
BC094407.1 - 2 - 3 225 573475 110
BY269662.1 - 2 - 3 296 110
Y17223.1 - 2 - 3 225 110

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5l

Panther Classification:

Molecular Function -

ATP synthase primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

ATP synthesis coupled proton transport
ATP metabolic process
hydrogen-exporting ATPase activity, phosphorylative mechanism
ATPase activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26470342 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26470347 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26470343 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026669 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000909 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048656 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067682 AK146520.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067682 BC086880.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067682 NM_013795.5 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2024021 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 458063 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 667379 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 255665 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181308 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50911 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 262596 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 671003 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 230918 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 274472 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 22656 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 109114 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64346 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 165047 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166730 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152740 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000909 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026669 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067682 AK146520.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067682 BC086880.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067682 NM_013795.5 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048656 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2024021 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002602 - -
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P75415 BC031384.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P75415 BC089555.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P75415 BY269662.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P75415 AK019350.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P75415 AK017092.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470348 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470378 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470340 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470336 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470380 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470343 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470364 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470394 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470396 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470377 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470353 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470366 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470352 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470376 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470383 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470390 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470398 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470375 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470369 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470373 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470337 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470381 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470387 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470350 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470345 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470363 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470392 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470334 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470389 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470356 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470341 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470371 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470354 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470384 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470359 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26470338 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01919427 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152740 - -
Whole Mouse Genome G4122A A_52_P75415 BC031384.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P75415 BC089555.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P75415 BY269662.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P75415 AK019350.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P75415 AK017092.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152740 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08162873 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08162871 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08162863 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26470343 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26470347 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26470342 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470354 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470304 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470267 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470350 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470289 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470364 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470262 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470261 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470366 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470297 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470265 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470294 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470293 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470352 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470264 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470301 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470278 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470274 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470260 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470272 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470359 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470281 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470292 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26470348 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470265 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470356 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08025897 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470357 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470350 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470275 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470261 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08060093 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470269 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470276 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470304 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470264 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470352 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07912085 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470268 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470366 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470348 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470272 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470354 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470364 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07781647 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470266 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470274 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470271 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26470273 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08025895 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10354740 - -
Affymetrix Mouse Gene 1.0 ST Array 10592986 - -
Affymetrix Mouse Gene 1.0 ST Array 10592985 - -
Affymetrix Mouse Gene 1.0 ST Array 10592987 - -
Affymetrix Mouse Gene 1.0 ST Array 10553841 - -
Affymetrix Mouse Gene 1.0 ST Array 10599334 - -
Affymetrix Mouse Gene 1.0 ST Array 10380822 - -
Affymetrix Mouse Gene 1.0 ST Array 10528663 - -
Affymetrix Mouse Gene 1.0 ST Array 10592984 - -
Affymetrix Mouse Gene 1.1 ST Array 10354739 - -
Affymetrix Mouse Gene 1.1 ST Array 10528662 - -
Affymetrix Mouse Gene 1.1 ST Array 10592983 - -
Affymetrix Mouse Gene 1.1 ST Array 10380821 - -
Affymetrix Mouse Gene 1.1 ST Array 10599333 - -
Affymetrix Mouse Gene 1.1 ST Array 10553840 - -
Mouse Genome 430 2.0 Array 1448203_at - -
Mouse Genome 430 2.0 Array 1448203_PM_at - -
Affymetrix Mouse Clariom S Array PSR0900018207.mm.1 NM_013795.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900018207.mm.1 NM_013795.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900018207.mm.1 TC0900002185.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900018200.mm.1 NM_013795.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900018200.mm.1 NM_013795.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900018200.mm.1 TC0900002185.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900018205.mm.1 NM_013795.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900018205.mm.1 NM_013795.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900018205.mm.1 TC0900002185.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900018198.mm.1 NM_013795.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900018198.mm.1 NM_013795.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900018198.mm.1 TC0900002185.mm.1 MTA TC
Mouse Expression Set 430 1448203_at - -
Mouse Expression Set 430 1448203_PM_at - -
Murine Genome U74v2 Set 93014_at - -
Murine Genome U74v2 Set 99678_PM_f_at - -
Murine Genome U74v2 Set 99678_f_at - -
Murine Genome U74v2 Set 93014_PM_at - -
Mouse Genome 430A 2.0 Array 1448203_PM_at - -
Mouse Genome 430A 2.0 Array 1448203_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 101500070 - -
MouseWG-6 v2 Expression BeadChip 103290671 - -
Illumina mouse-6 v1.0 expression beadchip 101500070 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2587859 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2705268 - -
mouse-6 v1.1 (Illumina) scl027425.3_8-S - -
Illumina Sentrix mouse-6.v1 bead array 101500070 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 101500070 - -
MouseRef-8 v2 Expression BeadChip ILMN_2587859 - -
MouseRef-8 v2 Expression BeadChip ILMN_2973897 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2705268 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2973897 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0