This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07297317_g1
See other UXT GE Assays ›
Gene Symbol:
Uxt
Gene Name:
ubiquitously expressed transcript
Gene Aliases:
0910002B17Rik
Chromosome Location:
Chr.X: 20951665 - 20962035 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07297317_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uxt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013840.3 NP_038868.2 2 - 3 151 78
GenBank mRNA AF092738.1 - 2 - 3 138 78
AK027917.1 - 2 - 3 152 78
AK086822.1 - 2 - 3 160 78
AK163983.1 - 2 - 3 191 78
BC029258.1 - 2 - 3 134 4483276 78
BC082294.1 - 1 - 2 120 30290324 78

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Uxt

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
microtubule cytoskeleton organization
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
centrosome organization
RNA polymerase II transcription corepressor activity
chromatin binding
beta-tubulin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0