This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07297574_m1
See other PSMG1 GE Assays ›
Gene Symbol:
Psmg1
Gene Name:
proteasome (prosome, macropain) assembly chaperone 1
Gene Aliases:
AW552102, Dscr2
Chromosome Location:
Chr.16: 95979935 - 95990903 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07297574_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Psmg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019537.2 NP_062410.1 4 - 5 473 86
GenBank mRNA AJ238270.1 - 4 - 5 475 86
AK042203.1 - 4 - 5 484 86
AK088118.1 - 4 - 5 486 86
BC032965.1 - 4 - 5 466 1196077 86
BC060285.1 - 4 - 5 487 6508084 86

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Psmg1

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

cerebellar granule cell precursor proliferation
proteasome assembly
proteasome core complex assembly
proteasome binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0