This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07297618_m1
See other SH3BGRL GE Assays ›
Gene Symbol:
Sh3bgrl
Gene Name:
SH3-binding domain glutamic acid-rich protein like
Gene Aliases:
1190008F14Rik
Chromosome Location:
Chr.X: 109095407 - 109162467 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07297618_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sh3bgrl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019989.2 NP_064373.1 3 - 4 497 100
GenBank mRNA AJ272504.1 - 3 - 4 367 100
AK049929.1 - 3 - 4 389 100
AK146588.1 - 3 - 4 405 100
AK151095.1 - 3 - 4 407 100
AK151501.1 - 3 - 4 367 100
AK165995.1 - 3 - 4 407 100
BC034161.1 - 3 - 4 451 4954986 100
BC043055.1 - 3 - 4 497 6418767 100

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sh3bgrl

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
SH3 domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline