This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07297803_m1
See other CABLES1 GE Assays ›
Gene Symbol:
Cables1
Gene Name:
CDK5 and Abl enzyme substrate 1
Gene Aliases:
Cables, ik3-1
Chromosome Location:
Chr.18: 11839274 - 11945627 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07297803_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cables1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001146287.1 NP_001139759.1 6 - 7 1754 101
NM_022021.2 NP_071304.2 5 - 6 1676 101
GenBank mRNA AF328140.1 - 5 - 6 1361 101
AK156273.1 - 6 - 7 1810 101
BC043661.1 - 5 - 6 878 101
BC145142.1 - 5 - 6 1660 9053693 101
BC150714.1 - 5 - 6 1660 9087625 101
BQ917292.1 - 3 - 4 361 101

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cables1

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
cell cycle
nervous system development
cell division
regulation of cell cycle
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0