This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07297897_s1
See other FAHD1 GE Assays ›
Gene Symbol:
Fahd1
Gene Name:
fumarylacetoacetate hydrolase domain containing 1
Gene Aliases:
1110025H10Rik, AV002085
Chromosome Location:
Chr.17: 24848896 - 24850302 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07297897_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fahd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023480.2 NP_075969.1 1 820 87
GenBank mRNA AB041600.1 - 1 654 87
AK003914.1 - 1 610 87
AK044920.1 - 1 619 87
AK141973.1 - 1 820 87
AK157292.1 - 1 819 87
BC011330.1 - 1 850 87
BC026949.1 - 1 806 4221757 87

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fahd1

Panther Classification:

Molecular Function -

hydrolase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
catalytic activity
oxaloacetate decarboxylase activity
hydrolase activity
acetylpyruvate hydrolase activity
fumarylpyruvate hydrolase activity
metal ion binding
acylpyruvate hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline