This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07298058_m1
See other GPR146 GE Assays ›
Gene Symbol:
Gpr146
Gene Name:
G protein-coupled receptor 146
Gene Aliases:
BC003323, PGR8
Chromosome Location:
Chr.5: 139377698 - 139396415 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07298058_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr146
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038703.2 NP_001033792.1 1 - 2 144 75
GenBank mRNA AK137454.1 - 1 - 2 146 75

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK137454.1, NM_001038703.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline