This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07298084_m1
See other TRIOBP GE Assays ›
Gene Symbol:
Triobp
Gene Name:
TRIO and F-actin binding protein
Gene Aliases:
AI428493, EST478828, Tara, mKIAA1662
Chromosome Location:
Chr.15: 78947724 - 79005869 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07298084_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Triobp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039155.1 NP_001034244.1 4 - 5 739 69
NM_001039156.1 NP_001034245.1 4 - 5 739 69
NM_138579.4 NP_613045.3 4 - 5 739 69
GenBank mRNA CN528900.1 - 4 - 5 556 69
DQ228000.1 - 4 - 5 739 69
DQ228001.2 - 4 - 5 739 69
DQ228002.2 - 4 - 5 739 69
DQ278602.1 - 3 - 4 358 69

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Triobp

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
regulation of actin cytoskeleton organization
cell division
positive regulation of substrate adhesion-dependent cell spreading
actin binding
ubiquitin protein ligase binding
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0