This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07298356_m1
See other POLR2E GE Assays ›
Gene Symbol:
Polr2e
Gene Name:
polymerase (RNA) II (DNA directed) polypeptide E
Gene Aliases:
2410021N14Rik, 25kDa, AW208866, RPB5, XAP4
Chromosome Location:
Chr.10: 80035953 - 80039659 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07298356_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr2e
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025554.2 NP_079830.2 1 - 2 78 79
GenBank mRNA AK010567.1 - 1 - 2 86 79
AK015823.1 - 1 - 2 76 79
AK088610.1 - 1 - 2 134 79
AK132109.1 - 1 - 2 133 79
AK144668.1 - 1 - 2 139 79
AK145201.1 - 1 - 2 142 79
AK155368.1 - 1 - 2 111 79
BC005599.1 - 1 - 2 131 79
BC026842.1 - 1 - 2 69 4210827 79
BC037681.1 - 1 - 2 133 79
BC045521.1 - 1 - 2 78 2647754 79

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK155368.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
transcription from RNA polymerase II promoter
RNA polymerase I activity
RNA polymerase II activity
RNA polymerase III activity
DNA binding
DNA-directed RNA polymerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0